Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB27B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004163.4 | NP_004154.2 | 3 | 423 | ||
XM_005266733.1 | XP_005266790.1 | 4 | 344 | |||
XM_006722518.1 | XP_006722581.1 | 3 | 701 | |||
XM_011526131.1 | XP_011524433.1 | 3 | 701 | |||
XM_017025913.1 | XP_016881402.1 | 6 | 1177 | |||
XM_017025914.1 | XP_016881403.1 | 5 | 572 | |||
GenBank mRNA | AF329499.1 | - | 3 | 423 | ||
AF498954.1 | - | 2 | 180 | |||
AK314115.1 | - | 3 | 701 | |||
BC027474.1 | - | 3 | 291 | 4520182 | ||
U57093.2 | - | 3 | 272 |