Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ITCH SIRNA Assays ›
          Gene Symbol:
          ITCH
          Gene Name:
          itchy E3 ubiquitin protein ligase
          Gene Aliases:
          ADMFD, AIF4, AIP4, NAPP1
          Chromosome Location:
          Chr.20: 34363235 - 34511773 on Build GRCh38
          UniGene ID:
          Hs.632272
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 120673

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ITCH
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001257137.2 NP_001244066.1 10 1119
          NM_001257138.2 NP_001244067.1 8 854
          NM_001324197.1 NP_001311126.1 10 1140
          NM_001324198.1 NP_001311127.1 9 1017
          NM_031483.6 NP_113671.3 9 996
          XM_005260570.1 XP_005260627.1 8 912
          XM_011529079.1 XP_011527381.1 8 932
          XM_017028089.1 XP_016883578.1 9 952
          XM_017028090.1 XP_016883579.1 10 1072
          XM_017028091.1 XP_016883580.1 8 867
          XM_017028092.1 XP_016883581.1 10 1072
          GenBank mRNA AB056663.2 - 9 906
          AB209747.1 - 1 13
          AF038564.1 - 4 414
          AF095745.1 - 7 789
          AK297809.1 - 9 814
          AK304090.1 - 8 813
          AK315212.1 - 9 955
          BC011571.1 - 10 1042 4560366

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_008395, NM_001243712, AK048303, AK037940, AK220164, and others.

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          positive regulation of T cell anergy
          ubiquitin-dependent protein catabolic process
          apoptotic process
          inflammatory response
          Notch signaling pathway
          protein ubiquitination
          negative regulation of NF-kappaB transcription factor activity
          negative regulation of type I interferon production
          protein K29-linked ubiquitination
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of apoptotic process
          innate immune response
          positive regulation of protein catabolic process
          negative regulation of JNK cascade
          negative regulation of alpha-beta T cell proliferation
          viral entry into host cell
          negative regulation of defense response to virus
          defense response to virus
          nucleotide-binding oligomerization domain containing signaling pathway
          protein K63-linked ubiquitination
          protein K48-linked ubiquitination
          regulation of protein deubiquitination
          ubiquitin-protein transferase activity
          protein binding
          ligase activity
          ubiquitin-like protein transferase activity
          ribonucleoprotein complex binding
          CXCR chemokine receptor binding
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline