Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UBE2D3 SIRNA Assays ›
          Gene Symbol:
          UBE2D3
          Gene Name:
          ubiquitin conjugating enzyme E2 D3
          Gene Aliases:
          E2(17)KB3, UBC4/5, UBCH5C
          Chromosome Location:
          Chr.4: 102794383 - 102868893 on Build GRCh38
          UniGene ID:
          Hs.518773
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 120768

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: UBE2D3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001300795.1 NP_001287724.1 4, 5 304
          NM_003340.6 NP_003331.1 4, 5 611
          NM_181886.3 NP_871615.1 4, 5 819
          NM_181887.2 NP_871616.1 4, 5 589
          NM_181888.3 NP_871617.1 4, 5 468
          NM_181889.2 NP_871618.1 4, 5 491
          NM_181890.2 NP_871619.1 4, 5 433
          NM_181891.2 NP_871620.1 4, 5 776
          NM_181892.3 NP_871621.1 3, 4 801
          NM_181893.2 NP_871622.1 3, 4 281
          XM_017008584.1 XP_016864073.1 3, 4 643
          GenBank mRNA AK095822.1 - 3, 4 716
          AK127304.1 - 2, 3 3224
          AK315775.1 - 4, 5 377
          AL833414.1 - 4, 5 395
          AU131013.1 - 4, 5 379
          BC003395.1 - 4, 5 291 3447673
          BC037894.2 - 4, 5 588 5275356
          BC066917.1 - 4, 5 322 4792462
          BG613555.1 - 4, 5 416
          BM541512.1 - 4, 5 573
          BQ428985.1 - 4, 5 286
          BU567649.1 - 4, 5 461
          BU570661.1 - 3, 4 236
          CR456916.1 - 3, 4 126
          DA335600.1 - 4, 5 369
          DA550074.1 - 4, 5 223
          DA557036.1 - 4, 5 538
          DA808523.1 - 4, 5 544
          DA965437.1 - 3, 4 287
          DB234695.1 - 3, 4 225
          HY109217.1 - 4, 5 433
          U39318.1 - 3, 4 171

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031237, XM_006233334, XM_006233336, XM_006233335, XM_006233338, and others.
          This siRNA also targets the following transcripts in Mouse: NM_025356, AK153410, AK009276, BC098220, AK165235, and others.

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          protein polyubiquitination
          DNA repair
          cellular protein modification process
          ubiquitin-dependent protein catabolic process
          protein monoubiquitination
          apoptotic process
          protein ubiquitination
          BMP signaling pathway
          TRIF-dependent toll-like receptor signaling pathway
          proteasome-mediated ubiquitin-dependent protein catabolic process
          protein autoubiquitination
          regulation of transcription from RNA polymerase II promoter in response to hypoxia
          protein K48-linked ubiquitination
          protein K11-linked ubiquitination
          positive regulation of protein targeting to mitochondrion
          ubiquitin-protein transferase activity
          protein binding
          ATP binding
          ubiquitin conjugating enzyme activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0