Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306085.1 | NP_001293014.1 | 2 | 371 | ||
NM_198531.4 | NP_940933.3 | 2 | 371 | |||
XM_011525963.2 | XP_011524265.1 | 3 | 537 | |||
XM_011525964.2 | XP_011524266.1 | 2 | 252 | |||
XM_011525965.2 | XP_011524267.1 | 3 | 533 | |||
XM_011525966.2 | XP_011524268.1 | 2 | 488 | |||
XM_017025726.1 | XP_016881215.1 | 3 | 537 | |||
XM_017025727.1 | XP_016881216.1 | 3 | 537 | |||
XM_017025728.1 | XP_016881217.1 | 3 | 537 | |||
XM_017025729.1 | XP_016881218.1 | 3 | 537 | |||
XM_017025730.1 | XP_016881219.1 | 3 | 537 | |||
XM_017025731.1 | XP_016881220.1 | 3 | 537 | |||
XM_017025732.1 | XP_016881221.1 | 3 | 537 | |||
XM_017025733.1 | XP_016881222.1 | 3 | 537 | |||
XM_017025734.1 | XP_016881223.1 | 3 | 537 | |||
XM_017025735.1 | XP_016881224.1 | 3 | 537 | |||
XM_017025736.1 | XP_016881225.1 | 3 | 537 | |||
XM_017025737.1 | XP_016881226.1 | 3 | 537 | |||
XM_017025742.1 | XP_016881231.1 | 3 | 537 | |||
GenBank mRNA | AB373110.1 | - | 1 | 79 | ||
AK097757.1 | - | 2 | 488 | |||
AK295981.1 | - | 2 | 238 | |||
AK308291.1 | - | 2 | 253 | |||
BC053561.1 | - | 2 | 156 | |||
BC125219.1 | - | 2 | 249 | 40123958 | ||
BC125220.1 | - | 2 | 249 | 40123959 | ||
HY028472.1 | - | 2 | 372 |