Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LTF SIRNA Assays ›
          Gene Symbol:
          LTF
          Gene Name:
          lactotransferrin
          Gene Aliases:
          GIG12, HEL110, HLF2, LF
          Chromosome Location:
          Chr.3: 46436005 - 46485234 on Build GRCh38
          UniGene ID:
          Hs.529517
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 120978

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LTF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199149.1 NP_001186078.1 3, 4 511
          NM_001321121.1 NP_001308050.1 3, 4 622
          NM_001321122.1 NP_001308051.1 6, 7 855
          NM_002343.5 NP_002334.2 3, 4 622
          GenBank mRNA AF332168.1 - 3, 4 329
          AK093852.1 - 6, 7 858
          AK290859.1 - 3, 4 367
          AK292813.1 - 3, 4 367
          AK298035.1 - 3, 4 511
          AK316330.1 - 3, 4 431
          AY137470.1 - 3, 4 329
          AY165046.1 - 3, 4 341
          AY178998.2 - 3, 4 329
          AY360320.1 - 3, 4 330
          AY493417.1 - 3, 4 439
          AY875691.1 - 3, 4 329
          BC015822.1 - 3, 4 371 4251222
          BC015823.1 - 3, 4 373 4294752
          BC022347.1 - 3, 4 367 4251058
          BP342557.1 - 3, 4 570
          CD722125.1 - 3, 4 355
          DQ522304.1 - 3, 4 329
          EU779935.1 - 3, 4 329
          EU794669.1 - 3, 4 364
          KF992208.1 - 3, 4 341
          KT006756.1 - 3, 4 329
          M83202.1 - 3, 4 336
          M83205.1 - 3, 4 315
          M93150.1 - 3, 4 346
          U07643.1 - 3, 4 426
          X52941.1 - 3, 4 323
          X53961.1 - 3, 4 623

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          regulation of cytokine production
          retina homeostasis
          innate immune response in mucosa
          transcription, DNA-templated
          proteolysis
          ion transport
          humoral immune response
          antibacterial humoral response
          antifungal humoral response
          negative regulation of lipopolysaccharide-mediated signaling pathway
          regulation of tumor necrosis factor production
          negative regulation of ATPase activity
          iron assimilation by chelation and transport
          positive regulation of osteoblast proliferation
          positive regulation of toll-like receptor 4 signaling pathway
          negative regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          cellular protein metabolic process
          negative regulation by host of viral process
          negative regulation of viral genome replication
          cell redox homeostasis
          positive regulation of osteoblast differentiation
          negative regulation of viral process
          positive regulation of NF-kappaB transcription factor activity
          bone morphogenesis
          positive regulation of protein serine/threonine kinase activity
          positive regulation of bone mineralization involved in bone maturation
          negative regulation of single-species biofilm formation in or on host organism
          positive regulation of chondrocyte proliferation
          negative regulation of tumor necrosis factor (ligand) superfamily member 11 production
          negative regulation of osteoclast development
          DNA binding
          serine-type endopeptidase activity
          iron ion binding
          protein binding
          heparin binding
          protein serine/threonine kinase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0