Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LTF SIRNA Assays ›
          Gene Symbol:
          LTF
          Gene Name:
          lactotransferrin
          Gene Aliases:
          GIG12, HEL110, HLF2, LF
          Chromosome Location:
          Chr.3: 46436005 - 46485234 on Build GRCh38
          UniGene ID:
          Hs.529517
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 120980

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LTF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199149.1 NP_001186078.1 7 921
          NM_001321121.1 NP_001308050.1 7 1032
          NM_001321122.1 NP_001308051.1 10 1265
          NM_002343.5 NP_002334.2 7 1032
          GenBank mRNA AF332168.1 - 7 739
          AK093852.1 - 10 1268
          AK290859.1 - 7 777
          AK292813.1 - 7 777
          AK298035.1 - 7 921
          AK303889.1 - 6 575
          AK316330.1 - 7 841
          AY137470.1 - 7 739
          AY165046.1 - 7 751
          AY178998.2 - 7 739
          AY360320.1 - 7 740
          AY493417.1 - 7 849
          AY875691.1 - 7 739
          BC015822.1 - 7 781 4251222
          BC015823.1 - 7 783 4294752
          BC022347.1 - 7 777 4251058
          DQ522304.1 - 7 739
          EU779935.1 - 7 739
          EU794669.1 - 7 774
          KF992208.1 - 7 751
          KT006756.1 - 7 739
          M83202.1 - 7 746
          M83205.1 - 7 725
          M93150.1 - 7 756
          U07643.1 - 7 836
          X52941.1 - 7 733
          X53961.1 - 7 1033

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          regulation of cytokine production
          retina homeostasis
          innate immune response in mucosa
          transcription, DNA-templated
          proteolysis
          ion transport
          humoral immune response
          antibacterial humoral response
          antifungal humoral response
          negative regulation of lipopolysaccharide-mediated signaling pathway
          regulation of tumor necrosis factor production
          negative regulation of ATPase activity
          iron assimilation by chelation and transport
          positive regulation of osteoblast proliferation
          positive regulation of toll-like receptor 4 signaling pathway
          negative regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          cellular protein metabolic process
          negative regulation by host of viral process
          negative regulation of viral genome replication
          cell redox homeostasis
          positive regulation of osteoblast differentiation
          negative regulation of viral process
          positive regulation of NF-kappaB transcription factor activity
          bone morphogenesis
          positive regulation of protein serine/threonine kinase activity
          positive regulation of bone mineralization involved in bone maturation
          negative regulation of single-species biofilm formation in or on host organism
          positive regulation of chondrocyte proliferation
          negative regulation of tumor necrosis factor (ligand) superfamily member 11 production
          negative regulation of osteoclast development
          DNA binding
          serine-type endopeptidase activity
          iron ion binding
          protein binding
          heparin binding
          protein serine/threonine kinase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0