Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004074.2 | NP_004065.1 | 2 | 425 | ||
GenBank mRNA | BC000501.1 | - | 418 | |||
BC063025.1 | - | 2 | 425 | 6018884 |