Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ELN SIRNA Assays ›
          Gene Symbol:
          ELN
          Gene Name:
          elastin
          Gene Aliases:
          SVAS, WBS, WS
          Chromosome Location:
          Chr.7: 74027772 - 74069907 on Build GRCh38
          UniGene ID:
          Hs.647061
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 121259

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ELN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000501.3 NP_000492.2 4, 5 591
          NM_001081752.2 NP_001075221.1 3, 4 561
          NM_001081753.2 NP_001075222.1 4, 5 591
          NM_001081754.2 NP_001075223.1 4, 5 591
          NM_001081755.2 NP_001075224.1 4, 5 591
          NM_001278912.1 NP_001265841.1 4, 5 591
          NM_001278914.1 NP_001265843.1 3, 4 561
          NM_001278915.1 NP_001265844.1 4, 5 591
          NM_001278916.1 NP_001265845.1 4, 5 591
          NM_001278917.1 NP_001265846.1 3, 4 561
          NM_001278918.1 NP_001265847.1 3, 4 561
          NM_001278939.1 NP_001265868.1 4, 5 591
          XM_005250188.1 XP_005250245.1 4, 5 266
          XM_011515868.1 XP_011514170.1 4, 5 277
          XM_011515869.1 XP_011514171.1 3, 4 206
          XM_011515871.1 XP_011514173.1 4, 5 277
          XM_011515872.1 XP_011514174.1 4, 5 277
          XM_011515873.1 XP_011514175.1 4, 5 277
          XM_011515876.1 XP_011514178.1 4, 5 277
          XM_011515877.1 XP_011514179.1 4, 5 277
          XM_017011813.1 XP_016867302.1 3, 4 177
          XM_017011814.1 XP_016867303.1 4, 5 277
          GenBank mRNA AB208942.1 - 3, 4 187
          AK075494.1 - 4, 5 268
          AK092232.1 - 3, 4 176
          AK094138.1 - 4, 5 207
          AK095990.1 - 4, 5 211
          AK122731.1 - 4, 5 205
          AK125511.1 - 3, 4 221
          AK130894.1 - 4, 5 253
          AK225659.1 - 4, 5 205
          AK304845.1 - 4, 5 591
          BC035570.1 - 4, 5 196
          BC050379.1 - 4, 5 209
          BC065566.1 - 4, 5 222 6153564
          BP284909.1 - 4, 5 283
          BX537939.1 - 4, 5 211
          BX538199.1 - 3, 4 193
          M36860.1 - 4, 5 241
          X52896.1 - 4, 5 240

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          respiratory gaseous exchange
          blood circulation
          cell proliferation
          organ morphogenesis
          extracellular matrix disassembly
          extracellular matrix organization
          extracellular matrix structural constituent
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0