Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000950.2 | NP_000941.1 | 5 | 446 | ||
NM_001142395.1 | NP_001135867.1 | 4 | 355 | |||
NM_001173489.1 | NP_001166960.1 | 5 | 465 | |||
NM_001173490.1 | NP_001166961.1 | 4 | 374 | |||
GenBank mRNA | AF009242.1 | - | 5 | 358 | ||
AK312904.1 | - | 4 | 294 | |||
BC030786.2 | - | 4 | 338 | |||
BC060833.1 | - | 4 | 294 | 30348133 | ||
DA245329.1 | - | 5 | 432 | |||
DA411839.1 | - | 5 | 404 | |||
DA417581.1 | - | 4 | 374 | |||
DB286340.1 | - | 4 | 357 |