Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FN1 SIRNA Assays ›
Gene Symbol:
FN1
Gene Name:
fibronectin 1
Gene Aliases:
CIG, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF
Chromosome Location:
Chr.2: 215360440 - 215436167 on Build GRCh38
UniGene ID:
Hs.203717
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
121356
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001306129.1
NP_001293058.1
1, 2
423
NM_001306130.1
NP_001293059.1
1, 2
423
NM_001306131.1
NP_001293060.1
1, 2
423
NM_001306132.1
NP_001293061.1
1, 2
423
NM_002026.3
NP_002017.1
1, 2
421
NM_054034.2
NP_473375.2
1, 2
421
NM_212474.2
NP_997639.1
1, 2
421
NM_212476.2
NP_997641.1
1, 2
421
NM_212478.2
NP_997643.1
1, 2
421
NM_212482.2
NP_997647.1
1, 2
421
XM_005246397.1
XP_005246454.1
1, 2
520
XM_005246398.1
XP_005246455.1
1, 2
520
XM_005246399.1
XP_005246456.1
1, 2
520
XM_005246401.1
XP_005246458.1
1, 2
520
XM_005246402.1
XP_005246459.1
1, 2
520
XM_005246403.1
XP_005246460.1
1, 2
520
XM_005246404.1
XP_005246461.1
1, 2
520
XM_005246405.1
XP_005246462.1
1, 2
520
XM_005246406.1
XP_005246463.1
1, 2
520
XM_005246407.1
XP_005246464.1
1, 2
520
XM_005246408.1
XP_005246465.1
1, 2
520
XM_005246409.1
XP_005246466.1
1, 2
520
XM_005246410.1
XP_005246467.1
1, 2
520
XM_005246411.1
XP_005246468.1
1, 2
520
XM_005246412.1
XP_005246469.1
1, 2
520
XM_005246414.1
XP_005246471.1
1, 2
520
XM_005246416.1
XP_005246473.1
1, 2
520
XM_017003692.1
XP_016859181.1
1, 2
520
XM_017003693.1
XP_016859182.1
1, 2
520
XM_017003694.1
XP_016859183.1
1, 2
520
XM_017003695.1
XP_016859184.1
1, 2
520
GenBank mRNA
AB191261.1
-
1, 2
421
AB209840.1
-
1, 2
413
AJ276395.1
-
1, 2
211
AJ535086.1
-
1, 2
211
AJ849445.1
-
1, 2
211
AL832202.1
-
1, 2
422
AL832771.1
-
1, 2
422
BC117176.1
-
1, 2
401
40125727
BC143754.1
-
1, 2
401
9052268
BC143763.1
-
1, 2
401
9052277
BX537590.1
-
1, 2
422
BX538017.1
-
1, 2
422
BX538018.1
-
1, 2
422
BX538045.1
-
1, 2
461
BX640608.1
-
1, 2
421
BX640731.1
-
1, 2
422
BX640802.1
-
1, 2
424
BX640803.1
-
1, 2
421
BX640875.1
-
1, 2
422
BX649182.1
-
1, 2
424
CR749281.1
-
1, 2
460
CR749316.1
-
1, 2
420
CR749317.1
-
1, 2
420
DA851446.1
-
1, 2
423
X02761.1
-
1, 2
75
Back To Top
More Information
Panther Classification:
Molecular Function -
extracellular matrix structural protein
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
cell activation
regulation of protein phosphorylation
platelet degranulation
response to ischemia
acute-phase response
cell-substrate junction assembly
cell adhesion
calcium-independent cell-matrix adhesion
positive regulation of cell proliferation
glial cell migration
regulation of cell shape
response to wounding
response to ozone
positive regulation of gene expression
positive regulation of peptidase activity
peptide cross-linking
extracellular matrix disassembly
extracellular matrix organization
integrin activation
substrate adhesion-dependent cell spreading
cellular response to vascular endothelial growth factor stimulus
endodermal cell differentiation
cellular response to platelet-derived growth factor stimulus
wound healing
negative regulation of apoptotic process
positive regulation of axon extension
positive regulation of fibroblast proliferation
leukocyte migration
positive regulation of chemotaxis
response to glucocorticoid
regulation of ERK1 and ERK2 cascade
cellular response to lipopolysaccharide
cellular response to mercury ion
cellular response to glucose stimulus
cellular response to interleukin-1
cellular response to prostaglandin E stimulus
cellular response to transforming growth factor beta stimulus
cellular response to BMP stimulus
positive regulation of substrate-dependent cell migration, cell attachment to substrate
negative regulation of transforming growth factor-beta secretion
protease binding
integrin binding
protein binding
collagen binding
heparin binding
peptidase activator activity
identical protein binding
mercury ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0