Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFRP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003014.3 | NP_003005.2 | 3 | 940 | ||
GenBank mRNA | AF026692.1 | - | 3 | 811 | ||
AK302357.1 | - | 3 | 862 | |||
AK303977.1 | - | 4 | 760 | |||
AK313029.1 | - | 3 | 830 | |||
BC047684.1 | - | 3 | 798 | 5742936 | ||
BC058911.1 | - | 3 | 818 | 5228231 | ||
BT019679.1 | - | 3 | 554 | |||
CR541731.1 | - | 3 | 554 | |||
CR541755.1 | - | 3 | 554 | |||
DQ420628.1 | - | 3 | 573 |
Set Membership: |
G-protein coupled receptor |