Search
Search

XRN2| Gene Symbol: XRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317960.1 | NP_001304889.1 | 4 | 642 | ||
| NM_012255.4 | NP_036387.2 | 4 | 482 | |||
| XM_017027722.1 | XP_016883211.1 | 4 | 637 | |||
| XM_017027723.1 | XP_016883212.1 | 4 | 411 | |||
| XM_017027724.1 | XP_016883213.1 | 4 | 637 | |||
| GenBank mRNA | AF152169.1 | - | 4 | 385 | ||
| AK001573.1 | - | 4 | 452 | |||
| AK302846.1 | - | 4 | 346 | |||
| AK303312.1 | - | 3 | 293 | |||
| AK314602.1 | - | 4 | 421 | |||
| AL136841.1 | - | 4 | 408 | |||
| AY382900.1 | - | 3 | 297 | |||
| CF994999.1 | - | 4 | 489 | |||