Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPPR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166252.1 | NP_001159724.1 | 6 | 2055 | ||
NM_014839.4 | NP_055654.2 | 7 | 2229 | |||
XM_011542498.2 | XP_011540800.1 | 7 | 2177 | |||
GenBank mRNA | AB007924.3 | - | 7 | 2225 | ||
AF357013.1 | - | 7 | 2002 | |||
AF541281.1 | - | 7 | 2236 | |||
AK291369.1 | - | 7 | 2029 | |||
AK300361.1 | - | 6 | 1799 | |||
AL834390.1 | - | 6 | 1742 | |||
AY304518.1 | - | 7 | 2177 | |||
BC041386.1 | - | 1 | 135 | |||
BC042963.1 | - | 7 | 2047 |