Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271718.1 | NP_001258647.1 | 4 | 708 | ||
NM_001271719.1 | NP_001258648.1 | 4 | 668 | |||
NM_001271720.1 | NP_001258649.1 | 4 | 663 | |||
NM_001271721.1 | NP_001258650.1 | 4 | 623 | |||
NM_001271722.1 | NP_001258651.1 | 2 | 554 | |||
NM_021114.3 | NP_066937.1 | 4 | 685 | |||
NR_073417.1 | 4 | 537 | ||||
NR_073418.1 | 3 | 575 | ||||
NR_073419.1 | 4 | 577 | ||||
XM_011534405.1 | XP_011532707.1 | 3 | 799 | |||
GenBank mRNA | BC022514.1 | - | 4 | 547 | 4812189 | |
BI520020.1 | - | 4 | 597 | |||
BI561149.1 | - | 4 | 715 | |||
BU853655.1 | - | 4 | 541 | |||
GU727622.1 | - | 4 | 547 | |||
X57655.1 | - | 4 | 546 |