Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321411.1 | NP_001308340.1 | 5 | 1121 | ||
NM_031917.2 | NP_114123.2 | 5 | 1197 | |||
XM_005260091.3 | XP_005260148.1 | 5 | 1364 | |||
XM_011528347.2 | XP_011526649.1 | 6 | 1226 | |||
XM_011528348.2 | XP_011526650.1 | 6 | 1400 | |||
XM_011528349.2 | XP_011526651.1 | 7 | 1859 | |||
XM_011528350.2 | XP_011526652.1 | 6 | 1711 | |||
XM_017027347.1 | XP_016882836.1 | 6 | 1384 | |||
GenBank mRNA | AB054064.1 | - | 5 | 1197 | ||
AF230330.1 | - | 5 | 1246 | |||
AY358276.1 | - | 5 | 1122 | |||
BC142632.1 | - | 5 | 1115 | 40147717 |