Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2U | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152489.1 | NP_689702.1 | 7 | 823 | ||
XM_005270498.4 | XP_005270555.1 | 7 | 1617 | |||
XM_006710379.3 | XP_006710442.1 | 7 | 758 | |||
XM_011540764.2 | XP_011539066.1 | 8 | 1558 | |||
XM_017000379.1 | XP_016855868.1 | 5 | 385 | |||
XM_017000380.1 | XP_016855869.1 | 6 | 429 | |||
XM_017000381.1 | XP_016855870.1 | 5 | 338 | |||
XM_017000382.1 | XP_016855871.1 | 4 | 251 | |||
XM_017000383.1 | XP_016855872.1 | 5 | 344 | |||
XM_017000384.1 | XP_016855873.1 | 4 | 242 | |||
GenBank mRNA | BC029895.1 | - | 7 | 823 | 5168406 |