Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENDOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004435.2 | NP_004426.2 | 2 | 727 | ||
XM_011518347.2 | XP_011516649.1 | 2 | 825 | |||
GenBank mRNA | AK130973.1 | - | 2 | 208 | ||
BC004922.2 | - | 2 | 602 | 3604703 | ||
BC016351.1 | - | 2 | 757 | 4080883 | ||
X79444.1 | - | 2 | 708 |