Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other OGG1 SIRNA Assays ›
          Gene Symbol:
          OGG1
          Gene Name:
          8-oxoguanine DNA glycosylase
          Gene Aliases:
          HMMH, HOGG1, MUTM, OGH1
          Chromosome Location:
          Chr.3: 9749944 - 9788246 on Build GRCh38
          UniGene ID:
          Hs.380271
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 122366

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: OGG1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002542.5 NP_002533.1 2 654
          NM_016819.3 NP_058212.1 2 654
          NM_016820.3 NP_058213.1 2 654
          NM_016821.2 NP_058214.1 2 654
          NM_016826.2 NP_058434.1 2 654
          NM_016827.2 NP_058436.1 2 654
          NM_016828.2 NP_058437.1 2 654
          NM_016829.2 NP_058438.1 2 654
          XM_011533760.1 XP_011532062.1 2 649
          XM_017006493.1 XP_016861982.1 2 649
          XM_017006494.1 XP_016861983.1 2 649
          XM_017006495.1 XP_016861984.1 2 649
          XM_017006496.1 XP_016861985.1 2 649
          XM_017006497.1 XP_016861986.1 2 649
          XM_017006498.1 XP_016861987.1 2 649
          XM_017006499.1 XP_016861988.1 2 649
          GenBank mRNA AB000410.1 - 2 579
          AB019528.1 - 1 27
          AB019529.1 - 1 27
          AB019530.1 - 1 70
          AB019531.1 - 1 70
          AF003595.1 - 2 367
          AF026691.1 - 2 518
          AK289858.1 - 2 607
          BC000657.2 - 2 608 3350168
          U88527.1 - 2 621
          U88620.1 - 2 621
          U96710.1 - 2 368
          Y11731.2 - 2 619
          Y11838.1 - 2 618
          Y13277.1 - 2 615

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          acute inflammatory response
          base-excision repair
          regulation of transcription, DNA-templated
          cellular response to DNA damage stimulus
          response to oxidative stress
          aging
          response to radiation
          response to light stimulus
          response to estradiol
          regulation of protein import into nucleus, translocation
          nucleotide-excision repair, DNA incision
          response to drug
          negative regulation of apoptotic process
          depurination
          depyrimidination
          response to ethanol
          response to folic acid
          cellular response to cadmium ion
          negative regulation of double-strand break repair via single-strand annealing
          damaged DNA binding
          endonuclease activity
          protein binding
          microtubule binding
          oxidized purine nucleobase lesion DNA N-glycosylase activity
          oxidized purine DNA binding
          8-oxo-7,8-dihydroguanine DNA N-glycosylase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0