Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FDFT1 SIRNA Assays ›
Gene Symbol:
FDFT1
Gene Name:
farnesyl-diphosphate farnesyltransferase 1
Gene Aliases:
DGPT, ERG9, SQS, SS
Chromosome Location:
Chr.8: 11795573 - 11839309 on Build GRCh38
UniGene ID:
Hs.593928
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
122386
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FDFT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001287742.1
NP_001274671.1
5, 6
1099
NM_001287743.1
NP_001274672.1
4, 5
901
NM_001287744.1
NP_001274673.1
3, 4
728
NM_001287745.1
NP_001274674.1
4, 5
807
NM_001287747.1
NP_001274676.1
3, 4
764
NM_001287748.1
NP_001274677.1
3, 4
426
NM_001287749.1
NP_001274678.1
3, 4
664
NM_001287750.1
NP_001274679.1
2, 3
666
NM_001287751.1
NP_001274680.1
2, 3
538
NM_004462.4
NP_004453.3
3, 4
610
XM_017013217.1
XP_016868706.1
5, 6
1391
GenBank mRNA
AK057726.1
-
3, 4
728
AK098682.1
-
188
AK293545.1
-
3, 4
664
AK297868.1
-
2, 3
609
AK300059.1
-
2, 3
428
AK300245.1
-
3, 4
387
AK311246.1
-
2, 3
377
AK311362.1
-
3, 4
796
AK315993.1
-
4, 5
677
AK316033.1
-
3, 4
631
AK316182.1
-
2, 3
626
AK316351.1
-
3, 4
391
AK316531.1
-
3, 4
376
AK316534.1
-
2, 3
538
BC003573.1
-
3, 4
457
3538137
BC009251.2
-
3, 4
457
2961623
BC029641.1
-
3, 4
420
4514761
BC034440.1
-
3, 4
420
BG752446.1
-
474
BI765996.1
-
5, 6
703
BT006704.1
-
3, 4
388
CR457033.1
-
3, 4
388
DC387479.1
-
3, 4
425
L06070.1
-
3, 4
397
L06105.1
-
3, 4
432
S76822.1
-
3, 4
453
X69141.1
-
3, 4
479
Back To Top
More Information
Panther Classification:
Molecular Function -
transferase
Gene Ontology Categories:
Function(s)
Process(es)
steroid biosynthetic process
cholesterol biosynthetic process
isoprenoid biosynthetic process
farnesyl diphosphate metabolic process
oxidation-reduction process
farnesyl-diphosphate farnesyltransferase activity
oxidoreductase activity
squalene synthase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-p89b7:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0