Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C2CD5 SIRNA Assays ›
Gene Symbol:
C2CD5
Gene Name:
C2 calcium dependent domain containing 5
Gene Aliases:
CDP138, KIAA0528
Chromosome Location:
Chr.12: 22448546 - 22544546 on Build GRCh38
UniGene ID:
Hs.271014
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
122711
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C2CD5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286173.1
NP_001273102.1
12
1423
NM_001286174.1
NP_001273103.1
10
1557
NM_001286175.1
NP_001273104.1
12
1456
NM_001286176.1
NP_001273105.1
11
1417
NM_001286177.1
NP_001273106.1
11
1390
NM_014802.2
NP_055617.1
11
1417
XM_005253538.2
XP_005253595.1
11
1352
XM_005253539.3
XP_005253596.1
3
109
XM_006719170.2
XP_006719233.1
11
1379
XM_017020264.1
XP_016875753.1
15
1709
XM_017020265.1
XP_016875754.1
15
1709
XM_017020266.1
XP_016875755.1
15
1709
XM_017020267.1
XP_016875756.1
13
1637
XM_017020268.1
XP_016875757.1
12
1571
XM_017020269.1
XP_016875758.1
15
1709
XM_017020270.1
XP_016875759.1
14
1517
XM_017020271.1
XP_016875760.1
14
1490
XM_017020272.1
XP_016875761.1
12
1418
XM_017020273.1
XP_016875762.1
12
1418
XM_017020274.1
XP_016875763.1
11
1379
XM_017020275.1
XP_016875764.1
14
1490
XM_017020276.1
XP_016875765.1
11
1352
XM_017020277.1
XP_016875766.1
11
1352
XM_017020278.1
XP_016875767.1
12
1418
XM_017020279.1
XP_016875768.1
7
432
GenBank mRNA
AB011100.3
-
10
1170
AK295862.1
-
12
1401
AK299353.1
-
10
1264
AK310753.1
-
12
1423
AY166851.1
-
11
1389
BC042498.1
-
10
1176
5174801
BC053885.1
-
6
641
BC117143.1
-
11
1253
40125694
BC143859.1
-
11
1280
9052375
BC143860.1
-
11
1253
9052376
BC143878.1
-
12
1286
9052394
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_029081, NM_001109688, NM_029897, NM_001286578, AK162114, and others.
Gene Ontology Categories:
Function(s)
Process(es)
vesicle fusion
positive regulation of glucose transport
positive regulation of vesicle fusion
cellular response to insulin stimulus
insulin receptor signaling pathway via phosphatidylinositol 3-kinase
intracellular protein transmembrane transport
protein localization to plasma membrane
positive regulation of protein targeting to membrane
positive regulation of glucose import in response to insulin stimulus
calcium ion binding
calcium-dependent phospholipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline