Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018982.4 | NP_061855.1 | 9 | 1052 | ||
NR_036639.1 | 9 | 1052 | ||||
NR_036640.1 | 8 | 832 | ||||
NR_135075.1 | 7 | 769 | ||||
XM_017001499.1 | XP_016856988.1 | 8 | 810 | |||
XM_017001500.1 | XP_016856989.1 | 8 | 873 | |||
XM_017001501.1 | XP_016856990.1 | 7 | 654 | |||
GenBank mRNA | AK000823.1 | - | 7 | 717 | ||
AK300240.1 | - | 8 | 914 | |||
AK315183.1 | - | 9 | 1030 | |||
AL713668.1 | - | 8 | 763 | |||
BC009674.1 | - | 4 | 365 | |||
BC064590.1 | - | 9 | 1018 | 5749036 |