Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UBR4 SIRNA Assays ›
Gene Symbol:
UBR4
Gene Name:
ubiquitin protein ligase E3 component n-recognin 4
Gene Aliases:
RBAF600, ZUBR1, p600
Chromosome Location:
Chr.1: 19074506 - 19210334 on Build GRCh38
UniGene ID:
Hs.148078
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
123234
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UBR4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_020765.2
NP_065816.2
48
7022
XM_011541108.2
XP_011539410.1
48
7122
XM_011541109.2
XP_011539411.1
48
7119
XM_011541110.2
XP_011539412.1
48
7122
XM_011541111.2
XP_011539413.1
48
7122
XM_011541112.2
XP_011539414.1
48
7107
XM_011541113.2
XP_011539415.1
48
7104
XM_011541114.2
XP_011539416.1
48
7122
XM_011541115.2
XP_011539417.1
48
7122
XM_011541116.2
XP_011539418.1
48
7122
XM_011541117.2
XP_011539419.1
48
7122
XM_011541118.2
XP_011539420.1
48
7122
XM_011541119.2
XP_011539421.1
48
7122
XM_011541120.2
XP_011539422.1
48
7122
XM_011541121.2
XP_011539423.1
48
7122
XM_017000822.1
XP_016856311.1
48
7122
XM_017000823.1
XP_016856312.1
48
7107
XM_017000824.1
XP_016856313.1
48
7122
XM_017000825.1
XP_016856314.1
48
7122
XM_017000826.1
XP_016856315.1
48
7104
XM_017000827.1
XP_016856316.1
48
7122
XM_017000828.1
XP_016856317.1
48
7104
XM_017000829.1
XP_016856318.1
48
7122
XM_017000830.1
XP_016856319.1
48
7104
GenBank mRNA
AB037728.1
-
31
4687
AF348492.1
-
48
7018
AK056218.1
-
8
1159
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
viral process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
protein binding
calmodulin binding
zinc ion binding
ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0