Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANKRD36 SIRNA Assays ›
          Gene Symbol:
          ANKRD36 ANKRD36B
          Gene Name:
          ankyrin repeat domain 36
          ankyrin repeat domain 36B
          Gene Aliases:
          UNQ2430
          KIAA1641
          Chromosome Location:
          Chr.2: 97113186 - 97264521 on Build GRCh38
          Chromosome Location:
          Chr.2: 97492663 - 97590245 on Build GRCh38
          UniGene ID:
          Hs.541894
          Hs.532921
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 123261

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANKRD36
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164315.1 NP_001157787.1 76 6125
          XM_011511130.1 XP_011509432.1 76 6357
          XM_017004009.1 XP_016859498.1 76 6448
          XM_017004010.1 XP_016859499.1 76 6354
          XM_017004011.1 XP_016859500.1 74 6255
          XM_017004012.1 XP_016859501.1 74 6255
          XM_017004013.1 XP_016859502.1 74 6255
          XM_017004014.1 XP_016859503.1 74 6255
          XM_017004015.1 XP_016859504.1 74 6255
          XM_017004016.1 XP_016859505.1 74 6255
          XM_017004017.1 XP_016859506.1 74 6255
          XM_017004018.1 XP_016859507.1 74 6255
          XM_017004019.1 XP_016859508.1 74 6255
          XM_017004020.1 XP_016859509.1 74 6255
          XM_017004021.1 XP_016859510.1 74 6255
          XM_017004022.1 XP_016859511.1 74 6255
          XM_017004023.1 XP_016859512.1 74 6255
          XM_017004024.1 XP_016859513.1 75 6219
          XM_017004025.1 XP_016859514.1 72 6153
          XM_017004026.1 XP_016859515.1 72 6153
          XM_017004027.1 XP_016859516.1 72 6153
          XM_017004028.1 XP_016859517.1 72 6153
          XM_017004029.1 XP_016859518.1 72 6153
          XM_017004030.1 XP_016859519.1 72 6153
          XM_017004031.1 XP_016859520.1 72 6153
          XM_017004032.1 XP_016859521.1 72 6153
          XM_017004033.1 XP_016859522.1 72 6153
          XM_017004034.1 XP_016859523.1 72 6153
          XM_017004035.1 XP_016859524.1 72 6153
          XM_017004036.1 XP_016859525.1 72 6153
          XM_017004037.1 XP_016859526.1 72 6153
          XM_017004038.1 XP_016859527.1 72 6153
          XM_017004039.1 XP_016859528.1 70 6051
          XM_017004040.1 XP_016859529.1 70 6051
          XM_017004041.1 XP_016859530.1 70 6051
          XM_017004042.1 XP_016859531.1 70 6051
          XM_017004043.1 XP_016859532.1 70 6051
          XM_017004044.1 XP_016859533.1 68 5949
          XM_017004045.1 XP_016859534.1 68 5949
          XM_017004046.1 XP_016859535.1 74 6112
          XM_017004047.1 XP_016859536.1 73 6044
          XM_017004048.1 XP_016859537.1 72 5971
          XM_017004049.1 XP_016859538.1 74 6146
          XM_017004050.1 XP_016859539.1 59 4550
          XM_017004051.1 XP_016859540.1 57 4764
          Gene Symbol: ANKRD36B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005263995.4 XP_005264052.1 44 4768
          XM_006712661.1 XP_006712724.1 44 4677
          GenBank mRNA AF432209.1 - 5 1201
          AK024934.1 - 6 509
          BC125132.1 - 6 509
          BC125133.1 - 6 509

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0