Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C11ORF49 SIRNA Assays ›
Gene Symbol:
C11orf49
Gene Name:
chromosome 11 open reading frame 49
Gene Aliases:
-
Chromosome Location:
Chr.11: 46936524 - 47164381 on Build GRCh38
UniGene ID:
Hs.368296
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
123383
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C11orf49
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001003676.2
NP_001003676.1
4, 5
490
NM_001003677.2
NP_001003677.1
4, 5
490
NM_001003678.2
NP_001003678.1
4, 5
490
NM_001278222.1
NP_001265151.1
3, 4
387
NM_024113.4
NP_077018.1
4, 5
490
NR_103471.1
5, 6
735
NR_103472.1
3, 4
359
XM_006718315.1
XP_006718378.1
4, 5
481
XM_011520364.2
XP_011518666.1
3, 4
552
XM_011520365.2
XP_011518667.1
4, 5
481
XM_017018276.1
XP_016873765.1
4, 5
481
XM_017018277.1
XP_016873766.1
3, 4
300
XM_017018278.1
XP_016873767.1
5, 6
664
XM_017018279.1
XP_016873768.1
5, 6
727
XM_017018280.1
XP_016873769.1
6, 7
1388
XM_017018281.1
XP_016873770.1
5, 6
1357
XM_017018282.1
XP_016873771.1
5, 6
664
GenBank mRNA
AK025863.1
-
4, 5
399
AK293609.1
-
3, 4
268
AK300813.1
-
3, 4
297
AK315639.1
-
4, 5
438
AK316318.1
-
5, 6
727
AL136575.1
-
4, 5
409
BC001860.2
-
4, 5
436
3534541
BC009455.1
-
4, 5
411
3535652
BC013988.2
-
4, 5
401
3349493
BC052291.1
-
4, 5
403
5936112
CR457351.1
-
4, 5
373
DB027437.1
-
4, 5
481
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0