Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAQR6 SIRNA Assays ›
Gene Symbol:
PAQR6
Gene Name:
progestin and adipoQ receptor family member 6
Gene Aliases:
PRdelta
Chromosome Location:
Chr.1: 156243321 - 156248488 on Build GRCh38
UniGene ID:
Hs.235873
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
123520
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PAQR6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001272104.1
NP_001259033.1
8
1155
NM_001272105.1
NP_001259034.1
9
1227
NM_001272106.1
NP_001259035.1
6
1121
NM_001272107.1
NP_001259036.1
7
1290
NM_001272108.1
NP_001259037.1
6
780
NM_001272109.1
NP_001259038.1
5
1045
NM_001272110.1
NP_001259039.1
7
1028
NM_001272111.1
NP_001259040.1
7
986
NM_001272112.1
NP_001259041.1
5
813
NM_001272113.1
NP_001259042.1
5
683
NM_024897.3
NP_079173.2
7
1361
NM_198406.2
NP_940798.1
8
1113
NR_073610.1
7
1037
XM_005245494.3
XP_005245551.1
6
1281
XM_006711546.2
XP_006711609.1
8
1124
XM_006711547.3
XP_006711610.1
8
1597
XM_006711548.3
XP_006711611.1
7
1479
XM_006711552.2
XP_006711615.1
6
1222
XM_006711553.3
XP_006711616.1
6
1264
XM_011510000.2
XP_011508302.1
7
1387
XM_017002385.1
XP_016857874.1
7
2755
GenBank mRNA
AF455045.1
-
7
1209
AF455046.1
-
5
990
AF455047.1
-
7
952
AK026325.1
-
4
553
AK054682.1
-
7
967
AK094346.1
-
7
1006
AK095143.1
-
8
1085
AK095197.1
-
7
1296
AK294143.1
-
7
965
AK295915.1
-
7
979
AK295916.1
-
6
1077
AK307441.1
-
6
817
AK316034.1
-
9
1166
AL834227.1
-
7
1181
AY424284.1
-
8
1034
BC058509.1
-
8
997
5770270
BC071623.1
-
3
274
FJ200490.1
-
4
482
FJ200491.1
-
5
544
FJ200492.1
-
5
609
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
steroid hormone mediated signaling pathway
response to steroid hormone
steroid hormone receptor activity
steroid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-zctgw:80/10.239.163.131:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0