Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198719.1 | NP_001185648.1 | 12 | 1704 | ||
NM_002893.3 | NP_002884.1 | 12 | 1716 | |||
GenBank mRNA | AK091911.1 | - | 12 | 1701 | ||
AK127332.1 | - | 3 | 3729 | |||
AK291567.1 | - | 12 | 1460 | |||
BC114500.1 | - | 12 | 1614 | 40083852 | ||
BC114501.1 | - | 12 | 1614 | 40083851 | ||
U35143.1 | - | 12 | 1619 | |||
X72841.1 | - | 12 | 1643 |