Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: BUD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159736.1 | NP_001153208.1 | 2 | 220 | ||
NM_032725.3 | NP_116114.1 | 2 | 220 | |||
XM_011543035.2 | XP_011541337.1 | 2 | 794 | |||
GenBank mRNA | AK057832.1 | - | 2 | 209 | ||
AK225164.1 | - | 2 | 209 | |||
AK289635.1 | - | 2 | 209 | |||
AK291111.1 | - | 2 | 209 | |||
AK311068.1 | - | 2 | 209 | |||
BC006350.2 | - | 2 | 210 | 4111572 | ||
DA042987.1 | - | 2 | 221 |