siRNA Details
Gene Symbol: CHM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000390.3 | NP_000381.1 | 15 | 3918 | ||
NM_001320959.1 | NP_001307888.1 | 15 | 3915 | |||
XM_017029243.1 | XP_016884732.1 | 15 | 4005 | |||
XM_017029244.1 | XP_016884733.1 | 15 | 3903 | |||
XM_017029245.1 | XP_016884734.1 | 15 | 4005 | |||
XM_017029246.1 | XP_016884735.1 | 15 | 3884 | |||
GenBank mRNA | AK095883.1 | - | 3 | 514 | ||
AY255796.1 | - | 1 | 326 | |||
BC065702.1 | - | 1 | 418 | |||
BC073987.1 | - | 1 | 711 | |||
BC105969.1 | - | 1 | 403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
