Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153450.2 | NP_703151.2 | 4 | 783 | ||
GenBank mRNA | BC037223.1 | - | 4 | 661 | 5197938 |