Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178454.4 | NP_848549.3 | 9 | 1037 | ||
XM_005270469.2 | XP_005270526.1 | 10 | 1095 | |||
XM_005270470.1 | XP_005270527.1 | 10 | 1218 | |||
XM_006710361.1 | XP_006710424.1 | 10 | 1055 | |||
XM_006710362.1 | XP_006710425.1 | 10 | 1028 | |||
XM_011540707.1 | XP_011539009.1 | 10 | 1036 | |||
XM_011540708.1 | XP_011539010.1 | 10 | 1116 | |||
XM_017000320.1 | XP_016855809.1 | 9 | 979 | |||
XM_017000321.1 | XP_016855810.1 | 9 | 838 | |||
GenBank mRNA | AK075350.1 | - | 5 | 483 | ||
AK309557.1 | - | 9 | 1258 | |||
AL833098.1 | - | 9 | 1405 | |||
AY336747.1 | - | 9 | 915 | |||
AY358492.1 | - | 9 | 900 | |||
BC047025.1 | - | 8 | 852 | 5491665 | ||
BC091509.1 | - | 10 | 1129 | 30513000 | ||
EF710624.1 | - | 7 | 780 |