Search
Search

PIFO| Gene Symbol: PIFO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181643.5 | NP_857594.2 | 3 | 340 | ||
| XM_005270472.1 | XP_005270529.1 | 3 | 449 | |||
| GenBank mRNA | AK074433.1 | - | 3 | 275 | ||
| BC029237.1 | - | 2 | 229 | |||
| BC050319.1 | - | 3 | 304 | 4837146 | ||
| BC068603.1 | - | 3 | 264 | |||
| BC093891.1 | - | 3 | 327 | 7939736 | ||
| BC101501.1 | - | 3 | 328 | 8069007 | ||
| HM237139.1 | - | 3 | 327 | |||