Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286451.1 | NP_001273380.1 | 4 | 469 | ||
NM_198527.3 | NP_940929.1 | 4 | 620 | |||
NR_104447.1 | 4 | 512 | ||||
GenBank mRNA | AK057584.1 | - | 1 | 1188 | ||
BC033794.1 | - | 4 | 620 | 5122164 | ||
BQ777217.1 | - | 4 | 419 | |||
CV573934.1 | - | 4 | 454 |