Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIGD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099668.1 | NP_001093138.1 | 2, 3 | 383 | ||
NM_001099669.1 | NP_001093139.1 | 2, 3 | 236 | |||
NM_014056.3 | NP_054775.2 | 2, 3 | 212 | |||
GenBank mRNA | AF077034.1 | - | 2, 3 | 164 | ||
AF145385.1 | - | 2, 3 | 151 | |||
AK225461.1 | - | 2, 3 | 136 | |||
AK225571.1 | - | 2, 3 | 167 | |||
AK311815.1 | - | 2, 3 | 167 | |||
AL110233.1 | - | 2, 3 | 182 | |||
BC000601.1 | - | 2, 3 | 150 | 3346355 | ||
BC009583.1 | - | 2, 3 | 143 | 4093421 | ||
BC009594.1 | - | 2, 3 | 149 | 4109190 | ||
BC070277.1 | - | 2, 3 | 165 | 4686431 | ||
BG708126.1 | - | 2, 3 | 217 | |||
BP208784.1 | - | 2, 3 | 382 | |||
BP370118.1 | - | 2, 3 | 158 | |||
CR457385.1 | - | 1, 2 | 90 | |||
DA428926.1 | - | 2, 3 | 378 |