Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF638 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014972.2 | NP_001014972.1 | 2 | 1327 | ||
NM_001252612.1 | NP_001239541.1 | 2 | 1653 | |||
NM_001252613.1 | NP_001239542.1 | 2 | 1337 | |||
NM_014497.4 | NP_055312.2 | 2 | 1337 | |||
XM_011532768.2 | XP_011531070.2 | 2 | 1423 | |||
XM_017003809.1 | XP_016859298.1 | 2 | 1428 | |||
XM_017003810.1 | XP_016859299.1 | 2 | 1338 | |||
XM_017003811.1 | XP_016859300.1 | 2 | 1423 | |||
XM_017003812.1 | XP_016859301.1 | 2 | 1423 | |||
GenBank mRNA | AF534078.1 | - | 1 | 1017 | ||
AK098006.1 | - | 1 | 124 | |||
AK291198.1 | - | 2 | 1301 | |||
AK293938.1 | - | 2 | 1358 | |||
AK309221.1 | - | 2 | 1378 | |||
BC024000.1 | - | 2 | 1296 | |||
BC064530.1 | - | 1 | 1061 | |||
BC083513.1 | - | 2 | 1296 | 5540321 | ||
BC143728.1 | - | 2 | 1290 | 9052242 | ||
BX537425.1 | - | 2 | 1301 | |||
CR749322.1 | - | 2 | 1650 | |||
D83032.1 | - | 2 | 1329 |