Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMCC1 SIRNA Assays ›
Gene Symbol:
TMCC1
Gene Name:
transmembrane and coiled-coil domain family 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 129647792 - 129893605 on Build GRCh38
UniGene ID:
Hs.477547
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
124831
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMCC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001017395.3
NP_001017395.2
4
1908
NM_001128224.2
NP_001121696.1
4
1309
NR_033361.1
2
1454
XM_006713542.3
XP_006713605.1
5
2021
XM_006713543.3
XP_006713606.1
5
2018
XM_006713544.1
XP_006713607.1
5
2146
XM_006713550.3
XP_006713613.1
5
1579
XM_006713552.3
XP_006713615.1
2
1201
XM_006713553.3
XP_006713616.1
2
2493
XM_011512572.2
XP_011510874.1
6
2181
XM_011512573.2
XP_011510875.1
5
2760
XM_011512576.2
XP_011510878.1
5
2014
XM_011512579.2
XP_011510881.1
4
1769
XM_011512580.2
XP_011510882.1
7
1906
XM_011512581.2
XP_011510883.1
6
1743
XM_011512582.1
XP_011510884.1
2
1535
XM_011512583.2
XP_011510885.1
4
1312
XM_011512584.2
XP_011510886.1
6
1631
XM_017005931.1
XP_016861420.1
5
1929
XM_017005932.1
XP_016861421.1
6
2168
XM_017005933.1
XP_016861422.1
4
1965
XM_017005934.1
XP_016861423.1
8
1979
XM_017005935.1
XP_016861424.1
9
2068
XM_017005936.1
XP_016861425.1
6
1742
XM_017005937.1
XP_016861426.1
6
1676
XM_017005938.1
XP_016861427.1
8
2042
XM_017005939.1
XP_016861428.1
7
1936
XM_017005940.1
XP_016861429.1
4
1314
XM_017005941.1
XP_016861430.1
4
1440
GenBank mRNA
AB018322.1
-
1
475
AK124671.1
-
2
1455
AK291448.1
-
4
1727
AK293408.1
-
2
991
BC012480.1
-
1
38
BC030619.2
-
2
1372
BC034284.1
-
1
58
BC039859.1
-
4
1815
5527623
BC047517.1
-
2
1372
BX640712.1
-
2
962
CR749206.1
-
2
1434
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0