Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TMCC1 SIRNA Assays ›
          Gene Symbol:
          TMCC1
          Gene Name:
          transmembrane and coiled-coil domain family 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 129647792 - 129893605 on Build GRCh38
          UniGene ID:
          Hs.477547
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 124832

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TMCC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001017395.3 NP_001017395.2 4, 5 1939
          NM_001128224.2 NP_001121696.1 4, 5 1340
          NR_033361.1 2, 3 1485
          XM_006713542.3 XP_006713605.1 5, 6 2052
          XM_006713543.3 XP_006713606.1 5, 6 2049
          XM_006713544.1 XP_006713607.1 5, 6 2177
          XM_006713550.3 XP_006713613.1 5, 6 1610
          XM_006713552.3 XP_006713615.1 2, 3 1232
          XM_006713553.3 XP_006713616.1 2, 3 2524
          XM_011512572.2 XP_011510874.1 6, 7 2212
          XM_011512573.2 XP_011510875.1 5, 6 2791
          XM_011512576.2 XP_011510878.1 5, 6 2045
          XM_011512579.2 XP_011510881.1 4, 5 1800
          XM_011512580.2 XP_011510882.1 7, 8 1937
          XM_011512581.2 XP_011510883.1 6, 7 1774
          XM_011512582.1 XP_011510884.1 2, 3 1566
          XM_011512583.2 XP_011510885.1 4, 5 1343
          XM_011512584.2 XP_011510886.1 6, 7 1662
          XM_017005931.1 XP_016861420.1 5, 6 1960
          XM_017005932.1 XP_016861421.1 6, 7 2199
          XM_017005933.1 XP_016861422.1 4, 5 1996
          XM_017005934.1 XP_016861423.1 8, 9 2010
          XM_017005935.1 XP_016861424.1 9, 10 2099
          XM_017005936.1 XP_016861425.1 6, 7 1773
          XM_017005937.1 XP_016861426.1 6, 7 1707
          XM_017005938.1 XP_016861427.1 8, 9 2073
          XM_017005939.1 XP_016861428.1 7, 8 1967
          XM_017005940.1 XP_016861429.1 4, 5 1345
          XM_017005941.1 XP_016861430.1 4, 5 1471
          GenBank mRNA AB018322.1 - 1, 2 506
          AK124671.1 - 2, 3 1486
          AK291448.1 - 4, 5 1758
          AK293408.1 - 2, 3 1022
          BC012480.1 - 1, 2 69
          BC030619.2 - 2, 3 1403
          BC034284.1 - 1, 2 89
          BC039859.1 - 4, 5 1846 5527623
          BC047517.1 - 2, 3 1403
          BX640712.1 - 2, 3 993
          CR749206.1 - 2, 3 1465

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0