Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015370.1 | NP_056185.1 | 2 | 453 | ||
XM_011530096.2 | XP_011528398.1 | 2 | 421 | |||
XM_017028741.1 | XP_016884230.1 | 2 | 376 | |||
GenBank mRNA | AL035364.1 | - | 2 | 453 | ||
BC074758.2 | - | 2 | 371 | 30915217 | ||
BC126262.1 | - | 2 | 375 | 8991978 | ||
BC126264.1 | - | 2 | 375 | 8991980 | ||
CR456405.1 | - | 2 | 340 |