Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABI3BP SIRNA Assays ›
Gene Symbol:
ABI3BP
Gene Name:
ABI family member 3 binding protein
Gene Aliases:
NESHBP, TARSH
Chromosome Location:
Chr.3: 100749335 - 100993508 on Build GRCh38
UniGene ID:
Hs.477015
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
124877
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABI3BP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015429.3
NP_056244.2
35
3970
XM_005247281.2
XP_005247338.1
68
6127
XM_005247282.2
XP_005247339.1
67
6085
XM_005247283.2
XP_005247340.1
67
6079
XM_005247284.2
XP_005247341.1
67
6070
XM_005247285.2
XP_005247342.1
67
6067
XM_005247287.2
XP_005247344.1
67
6061
XM_005247288.2
XP_005247345.1
67
6055
XM_005247289.2
XP_005247346.1
67
6052
XM_005247290.2
XP_005247347.1
67
6052
XM_005247291.2
XP_005247348.1
67
6049
XM_005247293.2
XP_005247350.1
66
6019
XM_005247294.2
XP_005247351.1
66
6010
XM_005247295.2
XP_005247352.1
66
5989
XM_005247296.2
XP_005247353.1
66
5986
XM_005247298.2
XP_005247355.1
66
5983
XM_005247299.3
XP_005247356.1
66
5995
XM_005247300.2
XP_005247357.1
65
5953
XM_005247301.2
XP_005247358.1
65
5926
XM_005247302.2
XP_005247359.1
65
5923
XM_005247303.2
XP_005247360.1
64
5866
XM_005247305.2
XP_005247362.1
63
5782
XM_005247306.2
XP_005247363.1
61
5719
XM_005247309.1
XP_005247366.1
66
5760
XM_006713569.2
XP_006713632.1
66
5974
XM_006713570.2
XP_006713633.1
65
5941
XM_006713571.2
XP_006713634.1
65
5911
XM_006713572.2
XP_006713635.1
65
5905
XM_011512639.1
XP_011510941.1
67
6067
XM_011512640.1
XP_011510942.1
67
6061
XM_011512641.1
XP_011510943.1
67
6052
XM_011512643.1
XP_011510945.1
66
6046
XM_011512644.1
XP_011510946.1
67
6043
XM_011512645.1
XP_011510947.1
67
6043
XM_011512646.1
XP_011510948.1
67
6034
XM_011512647.1
XP_011510949.1
66
5992
XM_011512648.1
XP_011510950.1
66
5989
XM_011512649.1
XP_011510951.1
66
5983
XM_011512651.1
XP_011510953.1
65
5926
XM_011512652.1
XP_011510954.1
65
5914
XM_011512653.1
XP_011510955.1
64
5905
XM_011512654.1
XP_011510956.1
65
5896
XM_011512656.1
XP_011510958.1
64
5830
XM_011512657.1
XP_011510959.1
64
5827
XM_011512658.1
XP_011510960.1
63
5815
XM_011512659.1
XP_011510961.1
63
5794
XM_011512660.1
XP_011510962.1
63
5764
XM_017006105.1
XP_016861594.1
67
6070
XM_017006106.1
XP_016861595.1
67
6052
XM_017006107.1
XP_016861596.1
65
5920
XM_017006108.1
XP_016861597.1
63
5830
GenBank mRNA
AB056106.1
-
18
2481
AK093861.1
-
18
2305
AL117664.1
-
10
1780
AL162038.1
-
5
1181
BC030221.1
-
35
3950
5205716
BX648726.1
-
44
4134
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cell-substrate adhesion
extracellular matrix organization
collagen binding
heparin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-8dfkb:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0