Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130420.1 | NP_001123892.1 | 8 | 780 | ||
NM_003075.3 | NP_003066.2 | 8 | 780 | |||
NM_139067.2 | NP_620706.1 | 8 | 780 | |||
XM_005269101.1 | XP_005269158.1 | 8 | 717 | |||
XM_005269102.1 | XP_005269159.1 | 8 | 717 | |||
XM_005269103.1 | XP_005269160.1 | 8 | 717 | |||
XM_005269104.1 | XP_005269161.1 | 8 | 717 | |||
XM_011538693.2 | XP_011536995.1 | 7 | 626 | |||
XM_017019884.1 | XP_016875373.1 | 8 | 717 | |||
XM_017019885.1 | XP_016875374.1 | 8 | 717 | |||
XM_017019886.1 | XP_016875375.1 | 8 | 717 | |||
XM_017019887.1 | XP_016875376.1 | 7 | 604 | |||
GenBank mRNA | AB209006.1 | - | 8 | 780 | ||
AB209293.1 | - | 8 | 689 | |||
AK315153.1 | - | 8 | 680 | |||
BC013045.1 | - | 8 | 706 | 4180388 | ||
BC026222.2 | - | 8 | 699 | 4395012 | ||
BT009924.1 | - | 8 | 674 | |||
U66616.1 | - | 8 | 696 |