Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SSUH2 SIRNA Assays ›
          Gene Symbol:
          SSUH2
          Gene Name:
          ssu-2 homolog (C. elegans)
          Gene Aliases:
          C3orf32, SSU-2, fls485
          Chromosome Location:
          Chr.3: 8619400 - 8745035 on Build GRCh38
          UniGene ID:
          Hs.736935
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 124919

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SSUH2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256748.1 NP_001243677.1 10, 11 906
          NM_001256749.1 NP_001243678.1 10, 11 927
          NM_015931.2 NP_057015.1 10, 11 1073
          NR_046358.1 11, 12 1154
          XM_011533774.1 XP_011532076.1 18, 19 2622
          XM_017006510.1 XP_016861999.1 11, 12 5387
          XM_017006511.1 XP_016862000.1 17, 18 2310
          XM_017006512.1 XP_016862001.1 17, 18 2479
          XM_017006513.1 XP_016862002.1 16, 17 2315
          XM_017006514.1 XP_016862003.1 16, 17 2221
          XM_017006515.1 XP_016862004.1 18, 19 3376
          XM_017006516.1 XP_016862005.1 13, 14 1737
          XM_017006517.1 XP_016862006.1 13, 14 1690
          XM_017006518.1 XP_016862007.1 18, 19 2601
          XM_017006519.1 XP_016862008.1 11, 12 5332
          XM_017006520.1 XP_016862009.1 11, 12 1683
          XM_017006521.1 XP_016862010.1 14, 15 1841
          XM_017006522.1 XP_016862011.1 14, 15 1732
          XM_017006523.1 XP_016862012.1 12, 13 1435
          XM_017006524.1 XP_016862013.1 13, 14 1673
          XM_017006525.1 XP_016862014.1 16, 17 2242
          XM_017006526.1 XP_016862015.1 11, 12 1321
          XM_017006527.1 XP_016862016.1 17, 18 2364
          XM_017006528.1 XP_016862017.1 11, 12 1360
          XM_017006529.1 XP_016862018.1 11, 12 1390
          XM_017006530.1 XP_016862019.1 13, 14 1823
          XM_017006531.1 XP_016862020.1 12, 13 1745
          XM_017006533.1 XP_016862022.1 17, 18 2397
          XM_017006534.1 XP_016862023.1 17, 18 2343
          XM_017006538.1 XP_016862027.1 15, 16 1940
          XM_017006539.1 XP_016862028.1 10, 11 1052
          XM_017006540.1 XP_016862029.1 11, 12 1231
          XM_017006541.1 XP_016862030.1 8, 9 873
          XM_017006542.1 XP_016862031.1 8, 9 851
          GenBank mRNA AB024705.1 - 10, 11 1046
          AK057972.1 - 11, 12 1150
          AK093066.1 - 16, 17 2103
          AK122938.1 - 15, 16 2671
          AK225925.1 - 11, 12 1150
          AK300175.1 - 10, 11 906
          BC052614.1 - 10, 11 910 6300057

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-dt4l6:80/100.66.76.4:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0