Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098426.1 | NP_001091896.1 | 3 | 735 | ||
XM_005257604.2 | XP_005257661.2 | 3 | 282 | |||
XM_017024967.1 | XP_016880456.1 | 3 | 1474 | |||
GenBank mRNA | AK300939.1 | - | 3 | 519 | ||
AK303951.1 | - | 3 | 321 | |||
AK307944.1 | - | 3 | 288 | |||
BC136322.1 | - | 4 | 604 | 9020309 | ||
BC142963.1 | - | 3 | 684 | 8860380 | ||
BM463728.1 | - | 3 | 232 | |||
U66618.1 | - | 3 | 678 |
Gene Symbol: PTCRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK130819.1 | - | 992 |