Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PBRM1 SIRNA Assays ›
Gene Symbol:
PBRM1
Gene Name:
polybromo 1
Gene Aliases:
BAF180, PB1
Chromosome Location:
Chr.3: 52545352 - 52685933 on Build GRCh38
UniGene ID:
Hs.189920
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
125129
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PBRM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_018313.4
NP_060783.3
3
332
XM_005265279.4
XP_005265336.1
4
560
XM_005265280.3
XP_005265337.1
3
377
XM_005265282.3
XP_005265339.1
4
519
XM_005265283.3
XP_005265340.1
3
374
XM_011533900.2
XP_011532202.1
6
726
XM_011533902.2
XP_011532204.1
5
658
XM_011533903.2
XP_011532205.1
5
663
XM_017006725.1
XP_016862214.1
5
606
XM_017006726.1
XP_016862215.1
6
716
XM_017006727.1
XP_016862216.1
6
661
XM_017006728.1
XP_016862217.1
5
657
XM_017006729.1
XP_016862218.1
5
1065
XM_017006730.1
XP_016862219.1
6
652
XM_017006731.1
XP_016862220.1
5
635
XM_017006732.1
XP_016862221.1
5
658
XM_017006733.1
XP_016862222.1
5
1052
XM_017006734.1
XP_016862223.1
5
625
XM_017006735.1
XP_016862224.1
5
1064
XM_017006736.1
XP_016862225.1
5
1065
XM_017006737.1
XP_016862226.1
4
939
XM_017006738.1
XP_016862227.1
4
560
XM_017006739.1
XP_016862228.1
4
936
XM_017006740.1
XP_016862229.1
4
477
XM_017006741.1
XP_016862230.1
5
640
XM_017006742.1
XP_016862231.1
5
663
XM_017006743.1
XP_016862232.1
5
1065
XM_017006744.1
XP_016862233.1
5
629
XM_017006745.1
XP_016862234.1
4
938
XM_017006746.1
XP_016862235.1
4
935
XM_017006747.1
XP_016862236.1
5
1066
XM_017006748.1
XP_016862237.1
4
938
XM_017006749.1
XP_016862238.1
4
504
XM_017006750.1
XP_016862239.1
3
392
XM_017006751.1
XP_016862240.1
5
640
XM_017006752.1
XP_016862241.1
5
1065
XM_017006753.1
XP_016862242.1
5
1064
XM_017006754.1
XP_016862243.1
5
658
XM_017006755.1
XP_016862244.1
5
640
XM_017006756.1
XP_016862245.1
5
640
XM_017006757.1
XP_016862246.1
3
363
XM_017006758.1
XP_016862247.1
3
494
XM_017006759.1
XP_016862248.1
5
640
XM_017006760.1
XP_016862249.1
5
658
XM_017006761.1
XP_016862250.1
5
667
XM_017006762.1
XP_016862251.1
5
658
XM_017006763.1
XP_016862252.1
6
717
XM_017006764.1
XP_016862253.1
5
640
XM_017006765.1
XP_016862254.1
4
519
XM_017006766.1
XP_016862255.1
5
647
GenBank mRNA
AF177387.1
-
2
221
AF197569.1
-
3
314
AF225870.1
-
2
221
AF225871.1
-
2
221
AF225872.1
-
2
221
AK056541.1
-
1
50
AY281068.1
-
2
220
BC115009.1
-
2
236
7962003
BC115010.1
-
2
237
BC115011.1
-
2
237
7962010
BC129934.1
-
2
221
40082629
BC129935.1
-
2
221
40082630
BE672944.1
-
3
332
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic nuclear division
negative regulation of cell proliferation
covalent chromatin modification
DNA binding
chromatin binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-tvqvm:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0