Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMCES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006109.1 | NP_001006109.1 | 3 | 470 | ||
NM_020187.2 | NP_064572.2 | 3 | 297 | |||
XM_005247636.3 | XP_005247693.1 | 3 | 482 | |||
XM_005247637.3 | XP_005247694.1 | 3 | 332 | |||
XM_017006877.1 | XP_016862366.1 | 3 | 314 | |||
GenBank mRNA | AF201934.1 | - | 3 | 310 | ||
AK025883.1 | - | 2 | 142 | |||
AK225264.1 | - | 3 | 292 | |||
AK297501.1 | - | 3 | 295 | |||
AK309891.1 | - | 3 | 294 | |||
BC009993.2 | - | 3 | 377 | 4130099 | ||
BC010125.2 | - | 3 | 260 | 3945258 | ||
BC050686.1 | - | 3 | 470 | 6155077 | ||
BC088363.1 | - | 3 | 294 | 30398247 |