Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMND5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022780.3 | NP_073617.1 | 5 | 1015 | ||
GenBank mRNA | AK022815.1 | - | 5 | 992 | ||
AK025448.1 | - | 1 | 96 | |||
BC012165.1 | - | 5 | 736 | 4636136 | ||
BC047668.1 | - | 5 | 945 | 6066798 |