siRNA Details
Gene Symbol: NABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031716.2 | NP_001026886.1 | 6 | 2275 | ||
NM_001254736.1 | NP_001241665.1 | 6 | 1874 | |||
NR_045622.1 | 7 | 1978 | ||||
NR_045623.1 | 7 | 2378 | ||||
GenBank mRNA | AK026486.1 | - | 7 | 2378 | ||
AL832659.1 | - | 6 | 1811 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
