Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003019.4 | NP_003010.4 | 6 | 673 | ||
XM_011540087.1 | XP_011538389.1 | 6 | 652 | |||
XM_011540088.2 | XP_011538390.1 | 5 | 624 | |||
GenBank mRNA | BC022318.1 | - | 6 | 674 | 4696173 | |
BI820265.1 | - | 5 | 643 | |||
CR541948.1 | - | 5 | 630 | |||
X65018.1 | - | 6 | 801 |