Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP13A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024524.3 | NP_078800.3 | 12 | 1578 | ||
XM_005269357.3 | XP_005269414.1 | 12 | 1599 | |||
XM_011513120.2 | XP_011511422.1 | 13 | 1700 | |||
XM_011513121.1 | XP_011511423.1 | 12 | 1578 | |||
XM_011513122.1 | XP_011511424.1 | 12 | 1272 | |||
XM_011513123.2 | XP_011511425.1 | 13 | 1705 | |||
XM_011513124.2 | XP_011511426.1 | 13 | 1705 | |||
GenBank mRNA | AJ306929.1 | - | 3 | 2102 | ||
DR005713.1 | - | 5 | 459 |