Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GTDC1 SIRNA Assays ›
Gene Symbol:
GTDC1
Gene Name:
glycosyltransferase like domain containing 1
Gene Aliases:
Hmat-Xa, mat-Xa
Chromosome Location:
Chr.2: 143945787 - 144333092 on Build GRCh38
UniGene ID:
Hs.44780
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
125489
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GTDC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001006636.3
NP_001006637.1
12
2580
NM_001164629.3
NP_001158101.1
12
2565
NM_001284233.1
NP_001271162.1
9
2244
NM_001284234.1
NP_001271163.1
10
2280
NM_001284238.1
NP_001271167.1
14
2779
NM_024659.4
NP_078935.2
10
2292
XM_005263774.2
XP_005263831.1
13
2664
XM_005263782.2
XP_005263839.1
13
2680
XM_005263784.2
XP_005263841.1
11
2539
XM_011511843.2
XP_011510145.1
13
2713
XM_011511855.2
XP_011510157.1
11
2657
XM_011511856.2
XP_011510158.1
11
2439
XM_017004917.1
XP_016860406.1
17
3335
XM_017004918.1
XP_016860407.1
15
2927
XM_017004919.1
XP_016860408.1
13
2813
XM_017004920.1
XP_016860409.1
15
5969
XM_017004921.1
XP_016860410.1
14
2762
XM_017004922.1
XP_016860411.1
14
2939
XM_017004923.1
XP_016860412.1
13
2727
XM_017004924.1
XP_016860413.1
15
2923
XM_017004925.1
XP_016860414.1
12
2585
XM_017004926.1
XP_016860415.1
10
2516
XM_017004927.1
XP_016860416.1
12
2547
XM_017004928.1
XP_016860417.1
14
2780
XM_017004929.1
XP_016860418.1
12
2517
XM_017004930.1
XP_016860419.1
12
3168
XM_017004931.1
XP_016860420.1
12
2581
XM_017004932.1
XP_016860421.1
12
2551
XM_017004933.1
XP_016860422.1
12
2533
XM_017004934.1
XP_016860423.1
13
5740
XM_017004935.1
XP_016860424.1
14
3610
XM_017004936.1
XP_016860425.1
13
2787
XM_017004937.1
XP_016860426.1
10
2523
XM_017004938.1
XP_016860427.1
9
2411
XM_017004939.1
XP_016860428.1
12
2571
XM_017004940.1
XP_016860429.1
9
2372
XM_017004941.1
XP_016860430.1
8
2261
XM_017004942.1
XP_016860431.1
11
2418
XM_017004943.1
XP_016860432.1
11
3084
XM_017004944.1
XP_016860433.1
10
2307
XM_017004945.1
XP_016860434.1
12
2525
XM_017004946.1
XP_016860435.1
10
2262
XM_017004947.1
XP_016860436.1
10
2945
XM_017004948.1
XP_016860437.1
11
5485
XM_017004949.1
XP_016860438.1
9
2161
XM_017004950.1
XP_016860439.1
11
2208
XM_017004951.1
XP_016860440.1
11
2163
XM_017004952.1
XP_016860441.1
11
2607
XM_017004953.1
XP_016860442.1
11
2179
XM_017004954.1
XP_016860443.1
12
2267
XM_017004955.1
XP_016860444.1
14
2615
XM_017004956.1
XP_016860445.1
8
2018
XM_017004957.1
XP_016860446.1
10
2064
GenBank mRNA
AB112440.1
-
1
337
AK021815.1
-
6
1756
AK091684.1
-
9
2248
AY281366.1
-
12
2580
BC017741.2
-
11
2332
4390876
BX647773.1
-
14
2718
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Process(es)
transferase activity, transferring glycosyl groups
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline