Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMKN SIRNA Assays ›
Gene Symbol:
DMKN
Gene Name:
dermokine
Gene Aliases:
UNQ729, ZD52F10
Chromosome Location:
Chr.19: 35497217 - 35513678 on Build GRCh38
UniGene ID:
Hs.417795
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
125799
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMKN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001035516.3
NP_001030593.1
3
160
NM_001126056.2
NP_001119528.2
15
1460
NM_001126059.2
NP_001119531.1
9
511
NM_001190347.1
NP_001177276.1
14
1412
NM_001308380.1
NP_001295309.1
10
574
NM_001308383.1
NP_001295312.1
8
481
NM_033317.4
NP_201574.3
13
1493
NR_033746.1
10
566
XM_006723477.1
XP_006723540.1
13
690
XM_006723484.1
XP_006723547.1
11
600
XM_006723489.1
XP_006723552.1
13
676
XM_006723493.2
XP_006723556.1
11
581
XM_006723494.2
XP_006723557.1
11
563
XM_006723503.2
XP_006723566.1
13
609
XM_011527494.2
XP_011525796.1
19
1699
XM_011527495.2
XP_011525797.1
19
1699
XM_011527496.2
XP_011525798.1
18
1663
XM_011527497.2
XP_011525799.1
18
1657
XM_011527498.2
XP_011525800.1
18
1657
XM_011527499.2
XP_011525801.1
18
1654
XM_011527500.2
XP_011525802.1
18
1645
XM_011527501.2
XP_011525803.1
18
1639
XM_011527502.2
XP_011525804.1
17
1609
XM_011527503.2
XP_011525805.1
17
1603
XM_011527504.2
XP_011525806.1
17
1585
XM_011527505.2
XP_011525807.1
16
1558
XM_011527506.2
XP_011525808.1
16
1549
XM_011527507.2
XP_011525809.1
15
1513
XM_011527508.2
XP_011525810.1
15
1498
XM_011527509.2
XP_011525811.1
14
1456
XM_011527510.2
XP_011525812.1
14
1453
XM_011527513.1
XP_011525815.1
10
549
XM_011527514.1
XP_011525816.1
9
489
XM_017027475.1
XP_016882964.1
11
594
XM_017027476.1
XP_016882965.1
9
517
XM_017027477.1
XP_016882966.1
9
401
GenBank mRNA
AF086315.1
-
3
151
AK096215.1
-
8
987
AL832080.1
-
3
161
AY622964.1
-
3
146
AY622965.1
-
13
1310
AY789695.1
-
13
1316
AY789698.1
-
11
545
AY789699.1
-
11
527
AY789700.1
-
10
485
AY789701.1
-
12
701
AY789702.1
-
10
491
AY789703.1
-
9
440
AY789704.1
-
10
509
AY789705.1
-
9
449
AY789706.1
-
8
398
AY789707.1
-
3
149
BC004493.2
-
11
483
BC011886.2
-
10
495
BC035311.1
-
13
1487
4752921
BM923917.1
-
10
495
BP326188.1
-
9
510
CN369399.1
-
8
420
EF158319.1
-
14
1235
EF158320.1
-
15
1287
EF158323.1
-
9
492
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cornified envelope assembly
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-29f27:80/100.66.78.233:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0