siRNA Details
Gene Symbol: SMARCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003079.4 | NP_003070.3 | 5, 6 | 369 | ||
GenBank mRNA | AK001532.1 | - | 5, 6 | 366 | ||
AK095047.1 | - | 5, 6 | 336 | |||
AK294218.1 | - | 2, 3 | 155 | |||
AK294666.1 | - | 3, 4 | 261 | |||
BC007082.1 | - | 5, 6 | 366 | 4076036 | ||
BC011017.1 | - | 5, 6 | 370 | 4287132 | ||
BC063700.1 | - | 5, 6 | 332 | 4540040 | ||
BC069196.1 | - | 5, 6 | 1026 | |||
BT007176.1 | - | 3, 4 | 245 | |||
EU327017.1 | - | 4, 5 | 231 | |||
EU327018.1 | - | 4, 5 | 231 | |||
EU327019.1 | - | 3, 4 | 187 | |||
EU327020.1 | - | 3, 4 | 187 | |||
EU327021.1 | - | 3, 4 | 250 |

More Information
Other Species Targeted

Panther Classification:
